Learn More
Sino Biological Influenza A H3N2 (A/swine/Colorado/A01203748/2012) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40689V08H100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/swine/Colorado/A01203748/2012 (H3N2)) hemagglutinin (AFU10042) (Met1-Ser525), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3649