Learn More
Sino Biological Influenza A H3N2 (A/District of Columbia/27/2023) Neuraminidase / NA Protein (ECD, His Tag)
Catalog No. 502993325
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/District of Columbia/27/2023(H3N2)) Neuraminidase / NA (EPI2990330) (Ile30-Thr469) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-299-3325 Supplier Sino Biological Supplier No. 41055V08B100
A DNA sequence encoding the Influenza A virus (A/District of Columbia/27/2023(H3N2)) Neuraminidase / NA (EPI2990330) (Ile30-Thr469) was expressed with a polyhistidine tag at the C-terminus.