Learn More
Sino Biological Influenza A H3N2 (A/Thailand/8/2022) Neuraminidase/NA Protein (ECD, His Tag)
Catalog No. 502993363
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Thailand/8/2022 (H3N2)) nucleoprotein (translated amino acids of EPI2178976) (Ile30-Thr469) with a vasodilator-stimulated phosphoprotein tetramerization domain at the C-terminus.
Catalog No. 50-299-3363 Supplier Sino Biological Supplier No. 41001V08B100
A DNA sequence encoding the Influenza A virus (A/Thailand/8/2022 (H3N2)) nucleoprotein (translated amino acids of EPI2178976) (Ile30-Thr469) with a vasodilator-stimulated phosphoprotein tetramerization domain at the C-terminus.