Learn More
Sino Biological Influenza A H3N2 (A/Babol/36/2005) Neuraminidase / NA Antibody, Rabbit PAb
Supplier:  Sino Biological 40017RP0120
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza A H3N2 (A/Babol/36/2005) Neuraminidase / NA (Catalog#40017-V07H; ACN50232.1; His36-Pro459). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-4310