Learn More
Sino Biological Influenza A H3N2 (A/Hong Kong/4801/2014) Neuraminidase / NA (His Tag)
Supplier:  Sino Biological 40569V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Hong Kong/4801/2014 (H3N2)) neuraminidase (translated amino acids of EPI675797) (His36-Ile469) was expressed with a vasodilator-stimulated phosphoprotein tetramerization domain at the N-terminusa and a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3699