Learn More
Sino Biological Influenza A H3N2 (A/Kansas/14/2017) Neuraminidase / NA Protein (ECD, His Tag)
Supplier:  Sino Biological 40766V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Kansas/14/2017 (H3N2)) neuraminidase (AVG71505.1) (Ile30-Ile469) was expressed with a vasodilator-stimulated phosphoprotein tetramerization domain at the N-terminus and a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3592