Learn More
Sino Biological Influenza A H3N2 (A/Australia/NHRC0001/2005) Neuraminidase / NA (His Tag)
Supplier:  Sino Biological 40745V07H100
Encompass_Preferred
A DNA sequence encoding the influenza A virus (A/Australia/NHRC0001/2005 (H3N2)) neuraminidase (AEG65596) (His36-Ile469), termed as NA, was fused with a N-terminal polyhistidine tag.
Catalog No. 50-255-3603