Learn More
Sino Biological Influenza A H3N2 (A/Hong Kong/CUHK19579/1998) Neuraminidase / NA (His Tag)
Supplier:  Sino Biological 40743V07H100
Encompass_Preferred
A DNA sequence encoding the influenza A virus (A/Hong Kong/CUHK19579/1998 (H3N2)) neuraminidase (ACF36533) (His36-Ile469), termed as NA, was fused with a N-terminal polyhistidine tag.
Catalog No. 50-255-3605