Learn More
Sino Biological Influenza A H3N2 Neuraminidase / NA (E119V mutation) (Active)
Supplier:  Sino Biological 40017VNAHC1600
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Babol/36/2005 (H3N2)) neuraminidase (ACN50232.1) (His 36-Pro 459) was expressed, the cell lysates are collected, and bio-activity was tested. There is an amino acid change from Glutamic to Valine (E119V mutation) in NA / Neuraminidase.
Catalog No. 50-255-3763