Learn More
Sino Biological H3N2 NP ELISA Pair Set 15PP (Sino Biological)
Catalog No. 502373184
Encompass_Preferred
Each vial contains 505 ng of recombinant Influenza A H3N2 (A/Hong Kong/1/1968) Nucleoprotein / NP. Reconstitute with 1 mL dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in dilution buffer, and a high standard of 15000 pg/mL is recommended.
Catalog No. 50-237-3184 Supplier Sino Biological Supplier No. SEK00615
Each vial contains 505 ng of recombinant Influenza A H3N2 (A/Hong Kong/1/1968) Nucleoprotein / NP. Reconstitute with 1 mL dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in dilution buffer, and a high standard of 15000 pg/mL is recommended.