Learn More
Sino Biological H3N2 NP MAb 20ug (Sino Biological)
Supplier:  Sino Biological 40208R06120
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Influenza A H3N2 (A/Hong Kong/1/1968) Nucleoprotein / NP Protein (Catalog# 40208-V08B; P22435.2; Met1-Asn498).
Catalog No. 50-237-3516