Learn More
Sino Biological Influenza A H3N2 (A/Panama/2007/99) Nucleoprotein / NP Protein (His Tag)
Supplier:  Sino Biological 40969V07B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Panama/2007/99 (H3N2)) Nucleoprotein / NP (EPI160205) (Met1-Asn498) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-240-8563