Learn More
Medchemexpress LLC HY-112219A 5mg Medchemexpress, H3R-IN-1 Hydrochloride CAS: Purity:>98%
SDP
Catalog No. 501969065
Encompass_Preferred
Medchemexpress, HY-112219A 5mg H3R-IN-1 HCl CAS: H3R-IN-1 Hydrochloride is a histamine receptor 3 (H3R) inverse agonist extracted from patent WO2013107336A1, compound example 2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9065 Supplier Medchemexpress LLC Supplier No. HY112219A5MG
Medchemexpress, HY-112219A 5mg H3R-IN-1 HCl CAS: H3R-IN-1 Hydrochloride is a histamine receptor 3 (H3R) inverse agonist extracted from patent WO2013107336A1, compound example 2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.