Learn More
Cayman Chemical H4 PosItIve Control 12.5nmol
SDP
Catalog No. 502893515
Encompass_Preferred
A PROTAC that drives FAK degradation binds to the FAK kinase domain (IC50 15.8 nM) and induces FAK degradation in A549 cells (DC50 1.3 nM) inhibits A549 cell migration and invasion at 100 nM reduces hepatic FAK levels in mice at 8 mg/kg
Catalog No. 50-289-3515 Supplier Cayman Chemical Supplier No. 60050712.5 NMOL
A PROTAC that drives FAK degradation binds to the FAK kinase domain (IC50 15.8 nM) and induces FAK degradation in A549 cells (DC50 1.3 nM) inhibits A549 cell migration and invasion at 100 nM reduces hepatic FAK levels in mice at 8 mg/kg