Learn More
Cayman Chemical H4 PosItIve Control 2.5nmol
SDP
Catalog No. 502893698
Encompass_Preferred
Oliogonucleotide recipes are optimized for BMS MIC. qPCR Enzyme positive controls and spike-in control are included. Includes 4 tubes Tube 1 5X Primers and probes for 4 pathogens and human control Tube 2 Synthetic positive controls Tube 3 Spike-in RPP30 control Tube 4 2X qPCR enzyme mastermix Organism Type Bacteria Gram- Virus dsDNA Multiplexed Reaction Panels STI Vaginal microbiota Colors Cy5 (HSV-1) FAM (Neisseria gonorrhoeae) HEX (RPP30) CalFluor-Red610 (Treponema pallidum)
Catalog No. 50-289-3698 Supplier Cayman Chemical Supplier No. 6005072.5 NMOL
Oliogonucleotide recipes are optimized for BMS MIC. qPCR Enzyme positive controls and spike-in control are included. Includes 4 tubes Tube 1 5X Primers and probes for 4 pathogens and human control Tube 2 Synthetic positive controls Tube 3 Spike-in RPP30 control Tube 4 2X qPCR enzyme mastermix Organism Type Bacteria Gram- Virus dsDNA Multiplexed Reaction Panels STI Vaginal microbiota Colors Cy5 (HSV-1) FAM (Neisseria gonorrhoeae) HEX (RPP30) CalFluor-Red610 (Treponema pallidum)