Learn More
Alkali Scientific Alkali Scientific™ H4000-H Hotplate, 7.5"x7.5", 115V
SDP
Supplier:  Alkali Scientific HP5000H
Encompass_Preferred
Ceramic work surface, 7.1 x 7.1 in. Stir speed from 60 to 1500 rpm Temperature control up to 380°C Three models: heat-stir, heat only or stir only Alkali hotplates, stirrers and hotplate-stirrers feature an exceptionally durable, chemical resistant white ceramic work surface. Their space-efficientdesign (8 X 9 in. footprint), makes them ideal for use on crowded benchtops and inside of bio-hoods. Advanced microprocessor controls with convenient turn knobs allow quick, precise adjustment and maintenance of speed and temperature. Safety indicator LED’s on the front panel indicate when the heating and/or stirring functions have been activated. With a square 7.1 in work surface, all three models are compatible with a wide variety of popular sizes of borosilicate glass beakers, flasks, bottles and other vessels. The hotplate/magnetic stirrer model (H4000-HS) comes complete with a support rod for mounting thermometers and temperature probes.
Catalog No. 50-302-9518