Learn More
Grainger Eaton HT800 Illuminated Pushbutton, 30.5 mm, Extended, Momentary, 120 V
Catalog No. 502773320
Encompass_Preferred
The HT800 series illuminated pushbutton is designed for robust performance in industrial environments. This 30.5 mm unit features a watertight and oiltight construction, ensuring reliable operation under challenging conditions. It is equipped with a clear lens and a full voltage LED light unit compatible with 120 Vac/dc, housed within a durable chrome bezel and plastic operator.
  • 30.5 mm diameter mounting size
  • Watertight and oiltight design for harsh environments
  • Momentary, extended actuator operation
  • Integrated LED, full voltage light unit at 120 Vac/dc
  • Chrome bezel with plastic operator
  • NEMA 3, 3R, 4, 4X, 12, and 13 enclosure ratings
  • CSA certified and UL listed
Catalog No. 50-277-3320 Supplier Grainger Supplier No. 13Z591
The HT800 series illuminated pushbutton is designed for robust performance in industrial environments. This 30.5 mm unit features a watertight and oiltight construction, ensuring reliable operation under challenging conditions. It is equipped with a clear lens and a full voltage LED light unit compatible with 120 Vac/dc, housed within a durable chrome bezel and plastic operator.
  • 30.5 mm diameter mounting size
  • Watertight and oiltight design for harsh environments
  • Momentary, extended actuator operation
  • Integrated LED, full voltage light unit at 120 Vac/dc
  • Chrome bezel with plastic operator
  • NEMA 3, 3R, 4, 4X, 12, and 13 enclosure ratings
  • CSA certified and UL listed