Learn More
Grainger Eaton Push Button Operator, Extended, Illuminated, 30 mm, 1 ea
Catalog No. 502770894
Encompass_Preferred
This HT800 series extended head operator is a 30.5 mm momentary pushbutton designed for industrial applications. It features an illuminated design with a clear head, providing versatile operation for various control systems. Engineered for durability, this operator includes corrosion-resistant properties and is suitable for use in demanding environments.
  • Corrosion resistant, oil and grease resistant, and watertight construction
  • Illuminated operator with full voltage LED lamp module
  • Screw clamp terminal connection
  • 30 mm panel mounting hole diameter
  • Compatible with NEMA 1, 2, 3, 3R, 4, 4X, 12, and 13 enclosures
  • Operational temperature range of 10°F to 140°F
  • 1NO-1NC contact type
Catalog No. 50-277-0894 Supplier Grainger Supplier No. 13Z598
This HT800 series extended head operator is a 30.5 mm momentary pushbutton designed for industrial applications. It features an illuminated design with a clear head, providing versatile operation for various control systems. Engineered for durability, this operator includes corrosion-resistant properties and is suitable for use in demanding environments.
  • Corrosion resistant, oil and grease resistant, and watertight construction
  • Illuminated operator with full voltage LED lamp module
  • Screw clamp terminal connection
  • 30 mm panel mounting hole diameter
  • Compatible with NEMA 1, 2, 3, 3R, 4, 4X, 12, and 13 enclosures
  • Operational temperature range of 10°F to 140°F
  • 1NO-1NC contact type