Learn More
Grainger Eaton HT800 Illuminated Pushbutton, Extended, Red, 30 mm, 1 ea
Catalog No. 502772207
Encompass_Preferred
The Eaton HT800 series extended pushbutton is a 30.5 mm illuminated operator designed for demanding industrial environments. This momentary, full-voltage LED device features a red lens and a chrome bezel, providing a reliable and durable solution for control applications. It is engineered to meet rigorous standards for watertight and oiltight performance.
  • Momentary action with extended head style
  • Illuminated red lens with 120 Vac/dc LED light unit
  • 30 mm mounting size for standard panel cutouts
  • Corrosion resistant design with chrome bezel
  • NEMA 3, 3R, 4, 4X, 12, and 13 enclosure ratings
  • Certified to UL and CSA standards
Catalog No. 50-277-2207 Supplier Grainger Supplier No. 13Z588
The Eaton HT800 series extended pushbutton is a 30.5 mm illuminated operator designed for demanding industrial environments. This momentary, full-voltage LED device features a red lens and a chrome bezel, providing a reliable and durable solution for control applications. It is engineered to meet rigorous standards for watertight and oiltight performance.
  • Momentary action with extended head style
  • Illuminated red lens with 120 Vac/dc LED light unit
  • 30 mm mounting size for standard panel cutouts
  • Corrosion resistant design with chrome bezel
  • NEMA 3, 3R, 4, 4X, 12, and 13 enclosure ratings
  • Certified to UL and CSA standards