Learn More
Diagenode Inc H4K5AC CHIP-SEQ GRADE 50UG
Catalog No. NC1850602
Encompass
H4K5ac Antibody - ChIP-seq Grade 50ug.Concentration 1.80 µg/µlSpecies reactivity Human, mouse, wide range expectedType Polyclonal
Catalog No. NC1850602 Supplier Diagenode Inc Supplier No. C15410025
H4K5ac Antibody - ChIP-seq Grade 50ug.Concentration 1.80 µg/µlSpecies reactivity Human, mouse, wide range expectedType Polyclonal