Learn More
Sino Biological Influenza A H4N2 (A/duck/Hunan/8-19/2009) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40390V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/duck/Hunan/8-19/2009(H4N2)) hemagglutinin (ADN78261.1) (Met1-Asp527), termed as HA, was expressed with a polyhistidine tag at the C-terminus
Catalog No. 50-255-3722