Learn More
Sino Biological Influenza A H4N6 (A/mallard/Ohio/657/2002) Hemagglutinin / HA Antibody (HRP), Mouse MAb
Supplier:  Sino Biological 11714MM03H20
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Influenza A H4N6 (A/mallard/Ohio/657/2002) Hemagglutinin / HA ( Catalog#11714-V08H; ABI47995.1; Met1-Ile528). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-255-4603