Learn More
Sino Biological Influenza A H4N6 Hemagglutinin / HA Antibody, Mouse MAb
Supplier:  Sino Biological 11714MM0520
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Influenza A virus H4N6 Hemagglutinin. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-9998