Learn More
Sino Biological Influenza A H4N6 Hemagglutinin / HA Antibody, Rabbit PAb, Antigen Affinity Purified
Supplier:  Sino Biological 11714RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant H4N6 (A/mallard/Ohio/657/2002) HA / Hemagglutinin (Catalog#11714-V08H; ABI47995.1; Met 1-Ile 528). H4N6 (A/mallard/Ohio/657/2002) HA / Hemagglutinin specific IgG was purified by H4N6 (A/mallard/Ohio/657/2002) HA / Hemagglutinin affinity chromatography.
Catalog No. 50-240-9649