Learn More
Sino Biological Influenza A H4N6 Hemagglutinin / HA Antibody, Rabbit PAb
Supplier:  Sino Biological 11714RP0120
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant H4N6 (A/mallard/Ohio/657/2002) HA / Hemagglutinin (Catalog#11714-V08H; ABI47995.1; Met 1-Ile 528). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8604