Learn More
Labtech Inc Recirculating Water Chille
Supplier:  Labtech Inc LW500
Encompass_Preferred
Recirculating Water Chiller by LabTech, cooling capacity of 500 Watt at 25˚C, 115V, 5C ~ 35˚C, model H50-500
Catalog No. NC1217691