Learn More
Alkali Scientific Alkali Scientific™ H5000-H MultiTherm Shaker with Heating ONLY
SDP
Catalog No. 503029506
Encompass_Preferred
Alkali Scientific™, H5000-H, MultiTherm Shaker with Heating Only, 115V The MultiTherm shaker is a temperature controlled vortexer, useful for a variety of molecular biology applications. Despite its modest 8 x 11 in. footprint, all popular tubes and plates are compatible, including as many as six 50ml tubes. Speed, time and temperature settings are continuously visible on the LCD, simultaneously showing both actual and selected values. Temperature control from 0°C to 100°C Exchangeable blocks, for tube sizes 0.2 to 50mL Precision contoured wells for uniform thermal transfer Two models: Heat/Cool or Heat only Sequence link function for linking or repeating programs (HC model only) Blocks sold separately
Catalog No. 50-302-9506 Supplier Alkali Scientific Supplier No. RS7106
Alkali Scientific™, H5000-H, MultiTherm Shaker with Heating Only, 115V The MultiTherm shaker is a temperature controlled vortexer, useful for a variety of molecular biology applications. Despite its modest 8 x 11 in. footprint, all popular tubes and plates are compatible, including as many as six 50ml tubes. Speed, time and temperature settings are continuously visible on the LCD, simultaneously showing both actual and selected values. Temperature control from 0°C to 100°C Exchangeable blocks, for tube sizes 0.2 to 50mL Precision contoured wells for uniform thermal transfer Two models: Heat/Cool or Heat only Sequence link function for linking or repeating programs (HC model only) Blocks sold separately