Learn More
Alkali Scientific H5000-HC Multi Therm Shaking V
SDP
Catalog No. 503029673
Encompass_Preferred
Temperature control from 0 to 100C Exchangeable blocks for tube sizes 0 2 to 50 ml Precision contoured wells for uniform thermal transfer Two models Heat/Cool or Heat only Sequence link function for linking or repeating programs (HC model only) Blocks sold separately The Alkali ScientificH5000-HC lab vortex mixer with heating and cooling is the economical choice for a variety of molecular biology applications Speed time and temperature settings are continuously visible on the LCD simultaneously showing both actual and selected values Integral over-temperature control ensures long life safety and sample integrity This model offer both heating and cooling and the capacity to shake samples at speeds up to 1500rpm The unique programmable feature allows for up to five heating and cooling cycling steps Heating ramp up rate is 5 degrees C per minute and cooling rate is 2 5 degrees C per minute Timer can be set from 1 minute to 99 hours or left on continuously A wide range of blocks are a
Catalog No. 50-302-9673 Supplier Alkali Scientific Supplier No. RS7105
Temperature control from 0 to 100C Exchangeable blocks for tube sizes 0 2 to 50 ml Precision contoured wells for uniform thermal transfer Two models Heat/Cool or Heat only Sequence link function for linking or repeating programs (HC model only) Blocks sold separately The Alkali ScientificH5000-HC lab vortex mixer with heating and cooling is the economical choice for a variety of molecular biology applications Speed time and temperature settings are continuously visible on the LCD simultaneously showing both actual and selected values Integral over-temperature control ensures long life safety and sample integrity This model offer both heating and cooling and the capacity to shake samples at speeds up to 1500rpm The unique programmable feature allows for up to five heating and cooling cycling steps Heating ramp up rate is 5 degrees C per minute and cooling rate is 2 5 degrees C per minute Timer can be set from 1 minute to 99 hours or left on continuously A wide range of blocks are a