Learn More
Genesee Scientific Corporation Benchmark Scientific H5000-HC MultiTherm Shaker, Temperature-Controlled, Heating & Cooling Functions, LCD Display
SDP
Catalog No. NC3584856
Encompass
The Benchmark Scientific H5000-HC MultiTherm Shaker offers precise temperature-controlled shaking with heating and cooling capabilities, making it ideal for various molecular biology applications. Its exchangeable blocks accommodate a range of tube sizes, while the LCD display provides real-time tracking of speed, time, and temperature. Equipped with integral over-temperature protection, this vortexer ensures sample integrity, setting it apart from standard shaking devices.
Catalog No. NC3584856 Supplier Genesee Scientific Corporation Supplier No. 31601
The Benchmark Scientific H5000-HC MultiTherm Shaker offers precise temperature-controlled shaking with heating and cooling capabilities, making it ideal for various molecular biology applications. Its exchangeable blocks accommodate a range of tube sizes, while the LCD display provides real-time tracking of speed, time, and temperature. Equipped with integral over-temperature protection, this vortexer ensures sample integrity, setting it apart from standard shaking devices.