Learn More
Alkali Scientific H5100-HCT Multi Therm Touch Sh
SDP
Catalog No. 503029544
Encompass_Preferred
The Alkali ScientificMulti-Therm Touch H5100-HCT lab vortex mixer with heating and cooling is the economical choice for a variety of molecular biology applications Large touch screen for simple and easy programming and viewing of status Advanced programming features include adjustable ramp rates preheat and clockwise/counterclockwise mixing Multiple block options for plates and tubes up to 50ml Broad temperature and speed range to meet a variety of applications Speed time and temperature settings are quickly entered on the large beautiful touch screen New pre-heat capabilities and bi-directional vortexing options Temperature ranges from 20 degrees below ambient to 105C In most typical laboratories the Multi-Therm Touch can keep samples cool at 4C The Alkali ScientificMultiTherm Touch has the capacity to shake samples at speeds up to 3000rpm Using the programmable touch screen up to five heating and cooling cycling steps are available Heating ramp up rate is 5 degrees C per minute an
Catalog No. 50-302-9544 Supplier Alkali Scientific Supplier No. RS5100HCT
The Alkali ScientificMulti-Therm Touch H5100-HCT lab vortex mixer with heating and cooling is the economical choice for a variety of molecular biology applications Large touch screen for simple and easy programming and viewing of status Advanced programming features include adjustable ramp rates preheat and clockwise/counterclockwise mixing Multiple block options for plates and tubes up to 50ml Broad temperature and speed range to meet a variety of applications Speed time and temperature settings are quickly entered on the large beautiful touch screen New pre-heat capabilities and bi-directional vortexing options Temperature ranges from 20 degrees below ambient to 105C In most typical laboratories the Multi-Therm Touch can keep samples cool at 4C The Alkali ScientificMultiTherm Touch has the capacity to shake samples at speeds up to 3000rpm Using the programmable touch screen up to five heating and cooling cycling steps are available Heating ramp up rate is 5 degrees C per minute an