Learn More
Grainger Eaton Illuminated Pushbutton, Momentary, Amber, 30.5 mm, 1NO
Catalog No. 502771143
Encompass_Preferred
The HT800 Series 30.5 mm illuminated momentary pushbutton is a heavy-duty operator designed for industrial applications requiring watertight and oiltight protection. Featuring a rugged metal construction and a chrome bezel, this device is engineered to withstand demanding environments while providing reliable switching performance.
  • Extended actuator with amber lens
  • 1NO contact configuration
  • 120 V AC/DC transformer light unit with long-lasting LED illumination
  • NEMA 3, 3R, 4, 4X, 12, and 13 rated for resistance to dust, water, oil, and corrosion
  • Clear transparent contact block housings for easy visual inspection of contacts
  • Stackable, color-coded contact blocks for flexible mounting and identification
  • Reliability ridge on movable contacts ensures solid electrical connection
Catalog No. 50-277-1143 Supplier Grainger Supplier No. 13Z627
The HT800 Series 30.5 mm illuminated momentary pushbutton is a heavy-duty operator designed for industrial applications requiring watertight and oiltight protection. Featuring a rugged metal construction and a chrome bezel, this device is engineered to withstand demanding environments while providing reliable switching performance.
  • Extended actuator with amber lens
  • 1NO contact configuration
  • 120 V AC/DC transformer light unit with long-lasting LED illumination
  • NEMA 3, 3R, 4, 4X, 12, and 13 rated for resistance to dust, water, oil, and corrosion
  • Clear transparent contact block housings for easy visual inspection of contacts
  • Stackable, color-coded contact blocks for flexible mounting and identification
  • Reliability ridge on movable contacts ensures solid electrical connection