Learn More
Biotang Inc Influenza A H5N1 (A/Indonesia/5/2005) Hemagglutinin / HA Protein (His Tag), 10ug, extracellular domain of Influenza A virus (ABW06108.1) hemagglutinin (Met 1-Gln 531) with cleavage site mutated (RESRRKKR- TETR, HA1+HA2, uncleaved)
Supplier:  Biotang Inc RPR9506
Encompass_Preferred
Influenza A H5N1 (A/Indonesia/5/2005) Hemagglutinin / HA Protein (His Tag), 10ug, extracellular domain of Influenza A virus (ABW06108.1) hemagglutinin (Met 1-Gln 531) with cleavage site mutated (RESRRKKR- TETR, HA1+HA2, uncleaved)
Catalog No. 50-751-5074