Learn More
Biotang Inc Influenza A H5N1 (A/Indonesia/5/2005) Hemagglutinin / HA Protein (His Tag), 10ug,extracellular domain of Influenza A virus (A/Indonesia/5/2005 (H5N1)) (ABW06108.1) hemagglutinin (Met 1-Gln 531) (Native, HA1+HA2, cleaved)
Supplier:  Biotang Inc RPR9507
Encompass_Preferred
Influenza A H5N1 (A/Indonesia/5/2005) Hemagglutinin / HA Protein (His Tag), 10ug,extracellular domain of Influenza A virus (A/Indonesia/5/2005 (H5N1)) (ABW06108.1) hemagglutinin (Met 1-Gln 531) (Native, HA1+HA2, cleaved)
Catalog No. 50-751-5073