Learn More
Biotang Inc Influenza A H5N1 (A/turkey/Turkey/1/2005) Hemagglutinin / HA Protein (His Tag) 10ug.the extracellular domain of Influenza A virus (ABD73284.1) hemagglutinin (Met 1-Gln 531) with cleavage site mutated (GERRRKKR - TETR, HA1+HA2, uncleaved)
Supplier:  Biotang Inc RPR9508
Encompass_Preferred
Influenza A H5N1 (A/turkey/Turkey/1/2005) Hemagglutinin / HA Protein (His Tag) 10ug.the extracellular domain of Influenza A virus (ABD73284.1) hemagglutinin (Met 1-Gln 531) with cleavage site mutated (GERRRKKR - TETR, HA1+HA2, uncleaved)
Catalog No. 50-751-5076