Learn More
Biotang Inc Influenza A H5N1 (A/Vietnam/1194/2004) Hemagglutinin / HA Protein (His Tag),10ug, extracellular domain of Influenza A virus (AAT73273.1) hemagglutinin (Met 1-Gln 531) with cleavage site mutated (RERRRKKR -TETR, HA1+HA2, uncleaved)
Supplier:  Biotang Inc RPR9510
Encompass_Preferred
Influenza A H5N1 (A/Vietnam/1194/2004) Hemagglutinin / HA Protein (His Tag),10ug, extracellular domain of Influenza A virus (AAT73273.1) hemagglutinin (Met 1-Gln 531) with cleavage site mutated (RERRRKKR -TETR, HA1+HA2, uncleaved)
Catalog No. 50-751-5078