Learn More
Sino Biological H5N1 HA ELISA Pair Set 15PP (Sino Biological)
Catalog No. 502371110
Encompass_Preferred
Each vial contains 300 ng of recombinant H5N1 hemagglutinin. Reconstitute with 1 mL dilution buffer. A seven-point standard curve usi ng 2-fold serial dilutions in dilution buffer, and a high standard of 8000 pg/mL is recommended.
Catalog No. 50-237-1110 Supplier Sino Biological Supplier No. SEKA00215
Each vial contains 300 ng of recombinant H5N1 hemagglutinin. Reconstitute with 1 mL dilution buffer. A seven-point standard curve usi ng 2-fold serial dilutions in dilution buffer, and a high standard of 8000 pg/mL is recommended.