Learn More
Sino Biological H5N1 HA MAb 20ul (Sino Biological)
Supplier:  Sino Biological 11048MM0420
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Influenza A virus H5N1 (Avian Flu) hemagglutinin extracellular domain. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-3005