Learn More
Sino Biological Influenza A H5N1 (Avian Flu) Hemagglutinin / HA Antibody (Biotin), Rabbit PAb
Supplier:  Sino Biological 11048RP02B100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza A virus H5N1 (Avian Flu) Hemagglutinin extracellular domain. Total IgG was purified by Protein A affinity chromatography and then biotinylated.
Catalog No. 50-255-4937