Learn More
Sino Biological Influenza A H5N1 (A/chicken/VietNam/NCVD-016/2008) Hemagglutinin (HA1 Subunit) Antibody, Rabbit PAb
Supplier:  Sino Biological 40158RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza A H5N1 (A/chicken/VietNam/NCVD-016/2008) Hemagglutinin (HA1 Subunit) ( Catalog#40158-V08H1; ACO07033.1; Met1-Arg343). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-0840