Learn More
Sino Biological Influenza H5N1 (A/chicken/VietNam/NCVD-016/2008) Hemagglutinin / HA1 Antibody, Rabbit PAb, Antigen Affinity Purified
Supplier:  Sino Biological 40158T62100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza H5N1 (A/chicken/VietNam/NCVD-016/2008) Hemagglutinin / HA1 ( Catalog#40158-V08H1; ACO07033.1; Met1-Arg343). Influenza H5N1 (A/chicken/VietNam/NCVD-016/2008) Hemagglutinin / HA1 specific IgG was purified by Influenza H5N1 (A/chicken/VietNam/NCVD-016/2008) Hemagglutinin / HA1 affinity chromatography.
Catalog No. 50-240-8334