Learn More
Sino Biological H5N1 HA PAb 20ul (Sino Biological)
Supplier:  Sino Biological 11700T6220
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza A H5N1 (A/Common magpie/Hong Kong/2256/2006) Hemagglutinin / HA ( Catalog#11700-V08H; ABJ96777.1; Met1-Glu530). Influenza A H5N1 (A/Common magpie/Hong Kong/2256/2006) Hemagglutinin / HA specific IgG was purified by Influenza A H5N1 (A/Common magpie/Hong Kong/2256/2006) Hemagglutinin / HA affinity chromatography.
Catalog No. 50-237-3464