Learn More
Sino Biological Influenza H5N1 (A/turkey/Turkey/1/2005) Hemagglutinin / HA Antibody, Rabbit PAb
Supplier:  Sino Biological 11061RP0120
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza H5N1 (A/turkey/Turkey/1/2005) Hemagglutinin / HA ( Catalog#11061-V08H1; ABD73284.1; Met1-Gln531). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2033