Learn More
Sino Biological Influenza H5N1 (A/Vietnam/1194/2004) Hemagglutinin / HA Antibody, Rabbit PAb
Supplier:  Sino Biological 11062RP0120
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza H5N1 (A/Vietnam/1194/2004) Hemagglutinin / HA ( Catalog#11062-V08H1; AAT73273.1; Met1-Gln531). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2185