Learn More
Sino Biological Influenza H5N1 (A/Egypt/N05056/2009) Hemagglutinin / HA Antibody, Rabbit PAb
Supplier:  Sino Biological 11702RP0120
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza H5N1 (A/Egypt/N05056/2009) Hemagglutinin / HA ( Catalog#11702-V08H; ACT15357.1; Met1-Gln530). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2095