Learn More
Sino Biological H5N1 HA PAb 20ul (Sino Biological)
Supplier:  Sino Biological 11710T6220
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza H5N1 (A/Cambodia/R0405050/2007) Hemagglutinin / HA ( Catalog#11710-V08H; ACI06178.1; Met1-Gln531). Influenza H5N1 (A/Cambodia/R0405050/2007) Hemagglutinin / HA specific IgG was purified by Influenza H5N1 (A/Cambodia/R0405050/2007) Hemagglutinin / HA affinity chromatography.
Catalog No. 50-237-0438