Learn More
Sino Biological H5N1 HA Protein 100ug (Sino Biological)
Supplier:  Sino Biological 40933V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus ((A/chicken/Ghana/AVL-763_21VIR7050-39/2021) (H5N1)) hemagglutinin (translated amino acids of EPI2415855) (Met1-Arg345), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-2296