Learn More
Sino Biological Influenza A H5N1 (A/Cambodia/X0123311/2013) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40698V08H100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Cambodia/X0123311/2013 (H5N1)) hemagglutinin (Met1-Ser526), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3641