Learn More
Sino Biological Influenza A H5N1 (A/Japanese white-eye/Hong Kong/1038/2006) Hemagglutinin Protein (HA1 Subunit) (His Tag), Biotinylated
Supplier:  Sino Biological 11694V08H1B20
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Japanese white-eye/Hong Kong/1038/2006 (H5N1)) hemagglutinin (ABJ96775.1) (Met1-Glu340), termed as HA1, was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-255-4607