Learn More
Sino Biological Influenza A H5N1 (A/Thailand/1(KAN-1)/2004) Neuraminidase / NA Protein (His Tag), Biotinylated
Supplier:  Sino Biological 40064V07HB20
Encompass_Preferred
A DNA sequence encoding the influenza A virus (A/Thailand/1(KAN-1)/2004(H5N1)) neuraminidase (His36-Ile469), termed as NA, was fused with a N-terminal polyhistidine tag. The expressed protein was biotinylated in vitro.
Catalog No. 50-255-3746