Learn More
Sino Biological Influenza A H5N1 Neuraminidase / NA (H255Y) (Active)
Supplier:  Sino Biological 11676VNAHC1300
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Anhui/1/2005 (H5N1)) neuraminidase (ABU94738.1) (Met 1-Lys 449) was expressed, the cell lysates are collected, and bio-activity was tested. There is an amino acid change from Histidine to Tyrosine (H255Y) in NA / Neuraminidase.
Catalog No. 50-255-4632